Features and Benefits
• Correctly folded & fully functional representation of 130 cancer proteins.
• Proprietary BCCP (biotin-carboxyl carrier protein) tagging technology ensures only fully functional cancer proteins are immobilized.
• Compatible with probing by DNA, proteins, kinases & small molecules.
Other Notes
Contains 130 recombinant human cancer proteins and experimental controls. Proteins are expressed in insect cells and arrayed onto a streptavidin-coated, low fluorescence glass slide (25 × 75.6 × 1 mm). The array consists of forty-eight, 4 × 4 sub-arrays, with proteins spotted in quadruplicate.
Legal Information
A Procognia proteomic technology product. CyDye is a registered trademark of GE Healthcare.
Panorama is a registered trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co.