Features and Benefits
• 259 correctly folded and fully functional proteins involved in signal transduction processes
• Proprietary BCCP (biotin-carboxyl carrier protein) tagging technology ensures only fully functional proteins are immobilized
• Probe with range of molecules including DNA, protein, kinases, or small molecules
Other Notes
Contains 259 human signaling proteins printed in quadruplicate plus specific controls. These proteins are intimately involved in many cell processes such as cell proliferation, cell differentiation and cell death. Proteins are expressed in Sf9 cells and arrayed onto a streptavidin-coated, low fluorescence glass slide (25 x 75.6 x 1 mm).
Legal Information
Panorama is a registered trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co.