antibody form
ascites fluid
clone
Ku15
mol wt
antigen mol wt 86 kDa
contains
15 mM sodium azide
species reactivity
human, monkey
application(s)
immunoblotting: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunohistochemistry (frozen sections): suitable
immunoprecipitation: suitable
indirect immunofluorescence: 1:10,000 using cultured human fibroblasts
microarray: suitable
isotype
IgG1
Gene Information
shipped in
dry ice
storage temp.
−20°C