Order Center
Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Prod. No. Only)
Supplier X Reference
SubStructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
L2034
Sigma
Lindane-UL-
14
C
≥95% (Radiochemical Purity), solid
Molecular Formula
C
6
H
6
Cl
6
Molecular Weight
290.83
CAS Number
180057-71-0
MDL number
MFCD00069503
Expand/Collapse All
Price and Availability
Descriptions
Packaging
screw cap bottle
Properties
assay
≥95% (Radiochemical Purity)
form
solid
extent of labeling
5-20 mCi per mmol
storage temp.
2-8°C
Safety
Hazard Codes
T+,R
Risk Statements
45-46-26/27/28-33
Safety Statements
53-26-36/37/39-45
RIDADR
UN 2910 7
WGK Germany
3
Related Categories
...
Pesticides
>
Acaricides
...
Pesticides
>
Labeled
...
Insecticides
>
Organochlorines
Scroll Up
Scroll Down
Related Information
MSDS
Certificate of Analysis
More Information
Links
Related Categories
Page Options
Printer Friendly View
Bulk Quote
Ask A Scientist
Email Page
Last 5 Products Viewed
L2034 (Sigma)
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.