|
| L2153 Sigma |
Latex beads, sulfate-modified polystyrene, fluorescent yellow greenaqueous suspension, 0.5 μm mean particle size |
|
|||||||
| Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. |