Analysis Note
Where reported, agglutination activity is expressed in μg/ml and is determined from serial dilutions in phosphate buffered saline, pH 6.8, of a 1 mg/ml solution. This activity is the lowest concentration to agglutinate a 2% suspension of human blood group A erythrocytes after 1 hour incubation at 25 °C.
Biochem/physiol Actions
HPA has anti-A human blood group specificity and has an affinity for terminal N-acetyl-α-D-galactosaminyl residues.1 Conjugates are prepared from affinity purified lectin.
Preparation Note
Prepared from peroxidase Type VI (P8375) using a modification of the method of O'Sullivan,2 which favors low molecular weight conjugates. Repurified by affinity chromatography after conjugation.
Unit Definition
One unit will form 1.0 mg purpurogallin from pyrogallol in 20 sec at pH 6.0 at 20 °C.
Physical form
Contains sodium chloride