Order Center
Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Prod. No. Only)
Supplier X Reference
SubStructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
LAA10
Sigma
L
-Amino acids
Expand/Collapse All
Price and Availability
Descriptions
Other Notes
10 Essential Amino Acids, 1 g of each:
• A5131
L
-Arginine Hydrochloride
• H8125
L
-Histidine Hydrochloride
• I2752
L
-Isoleucine
• L8000
L
-Leucine
• L5626
L
-Lysine Hydrochloride
• M9625
L
-Methionine
• P2126
L
-Phenylalanine
• T8625
L
-Threonine
• T0254
L
-Tryptophan
• V0500
L
-Valine
Related Categories
...
A
>
AM to AQ
...
Amino Acids
>
Amino Acids
...
Amino Acids, Peptides and Proteins
>
Amino Acids
Scroll Up
Scroll Down
Related Information
MSDS
Certificate of Analysis
More Information
Links
Similar Products
Related Categories
Page Options
Printer Friendly View
Bulk Quote
Ask A Scientist
Email Page
Last 5 Products Viewed
LAA10 (Sigma)
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.