Immunogen
recombinant human MCP-2 expressed in E. coli.
Specificity
The antibody may be used for neutralization, localization and detection of human MCP-2. It will not neutralize recombinant human MCP-1 or MCP-3, or recombinant mouse JE. The antibody shows <40% cross-reactivity with recombinant human HCC-1, <10% cross-reactivity with recombinant human eotaxin, and <5% cross-reactivity with recombinant human MCP-1.
Physical form
Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.