antibody form
affinity isolated antibody
form
buffered aqueous glycerol solution
species reactivity
human
concentration
~1 mg/mL
application(s)
immunoblotting (chemiluminescent): 1:2,000 using concentrated cell culture medium from a stimulated human cell line (M 2928)
immunoblotting: 1:1,000 using concentrated cell culture medium from a stimulated human cell line (M 2928)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
immunoprecipitation: suitable
indirect ELISA: suitable
Gene Information
shipped in
wet ice
storage temp.
−20°C