![]() |
Synonym
β
2
2
Decarboxycysteine
Thioethanolamine
Molecular Formula
NH2CH2CH2SH
Molecular Weight
77.15
CAS Number
Beilstein Registry Number
635649
MDL number
PubChem Substance ID
EG/EC Number
200-463-0
| ||||||||