Biochem/physiol Actions
Induces gastric, liver, kidney and lung cancer in mice and rats.
Caution
OSHA's Safety and Health Standard 29 CFR 1910 does not apply to a solution or mixture containing less than 1%. Injecting any compatible solvent permits preparation of any desired strength solution without exposure.
Packaging
Packaged in a 100 mL serum bottle with butyl rubber stopper and aluminum tear seal.
Reconstitution
Dissolving the contents in 100 mL of solvent yields a 1% solution.
Legal Information
Isopac is a registered trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co.