![]() |
Synonym
Ethanedioic acid
Molecular Formula
HO2CCO2H · 2H2O
Molecular Weight
126.07
CAS Number
Beilstein Registry Number
3679436
MDL number
PubChem Substance ID
EG/EC Number
205-634-3
| ||||||||