Order Center
Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Prod. No. Only)
Supplier X Reference
SubStructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
O6378
Sigma
Oligothymidylic acid d(pT)
12-18
ammonium salt
lyophilized powder
Synonym
d(pT)
12
-
18
MDL number
MFCD00131759
Expand/Collapse All
Price and Availability
Descriptions
Preparation Note
Prepared by automated DNA synthesis.
Unit Definition
One A
265
unit = approx. 20 μg oligonucleotide
Properties
assay
≥90% (HPLC)
form
lyophilized powder
storage temp.
−20°C
Safety
WGK Germany
3
Related Categories
...
Nucleosides, Nucleotides, Oligonucleotides
>
Synthetic Oligonucleotides and Related Reagents
Scroll Up
Scroll Down
Related Information
MSDS
Specification Sheet
Certificate of Analysis
More Information
Links
Related Categories
Page Options
Printer Friendly View
Bulk Quote
Ask A Scientist
Email Page
Last 5 Products Viewed
O6378 (Sigma)
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.