General description
These models offer power, versatility and ease-of-use. All models operate in constant voltage, current or power operating modes with automatic crossover between modes. Each is compact and lightweight for easy movement. All models feature a large LCD screen to display complete run information on one screen. At each step in programming, the power supply presents the options to the user in clear, concise language. The user simply presses the “smart key” corresponding to their choice. This information is retained when the unit turned off. These units can store up to 20 ten step programs in permanent memory.
Legal Information
EC Savant is a registered trademark of ThermoSavant.