Order Center
Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Prod. No. Only)
Supplier X Reference
SubStructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
P0431
Sigma
Proteose Peptone
Enzymatic hydrolysate
EG/EC Number
293-426-3
Expand/Collapse All
Price and Availability
Descriptions
Application
Recommended for use in antibiotic production and growth of fastidious microorganisms. Originally adapted for use in culture media for the production of bacterial toxins; a satisfactory nutrient for general culture needs.
Properties
nitrogen analysis
10.0-15.5% total
Safety
WGK Germany
3
Related Categories
...
Essential Microbiology Reagents
>
Media Components
...
Media Components
>
Protein Hydrolysates
...
Base Ingredients
>
Protein Sources (Peptones/Extracts)
Scroll Up
Scroll Down
Related Information
MSDS
Specification Sheet
Certificate of Analysis
Certificate of Origin
More Information
Links
Related Categories
Page Options
Printer Friendly View
Ask A Scientist
Email Page
Last 5 Products Viewed
P0431 (Sigma)
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.