General description
An all purpose analog pump designed to handle laboratory fluid transfer tasks. Economical and simple to operate, analog setting offers infinite flowrate adjustment. The chemically-resistant pump head (molded of glass filled Noryl®) has three stainless steel rollers and a Plexiglas safety cover.
• Standard Fluid Transfer: 2 year warranty
• Variable Flow from 1 mL/min to 3450 mL/min
• Analog single-turn speed control
• Lighted on/off switch. The on/off switch lets you maintain your flow rate setting when pump is turned off
• Reversible for purging of transfer lines
• 1/10 hp reversible motor has soft start and back EMF for 3% speed control
• Stackable painted steel housing is IP22
• 230 V models meet CE regulations for safety and noise immunity, 115 V models meet UL 508, IEC 1010 and cUL
All drives come complete with pump head and 4 ft silicone tubing (size 17). Start/stop operation by an open contact closure or open collector driver. Order foot switch model 73-750-000; (Remote models only). Operating temperature 0-40 °C.
Legal Information
Manostat is a registered trademark of Cole-Parmer Instrument Co.
Noryl is a registered trademark of General Electric Co.