Immunogen
recombinant N-terminal portion (amino acids 1-205) of mouse ΔNp63 isoform.1
Specificity
By immunoblotting, 3 bands are visible at approx. 50, 60, and 70 kDa using HaCaT cells, and multiple bands of approx. 44, 60, 80, and 90-95 kDa in other preparations.
Legal Information
This product is licensed for RESEARCH USE ONLY under U.S. Patents Nos. 6,946,256 and 7,030,227 owned by Harvard College. Other uses of the Sigma product, for example in diagnostics or therapeutics, are outside the scope of the license and are not authorized. For products licensed for diagnostic use, consult AsymmetRx, Inc. by telephone at 203-228-0209 or by email at pmckeon@asymmetrix.com