General description
Protein Molecular Mass (kDa) Phosphorylation Sites*
β-galactosidase 116 None
Phosphorylase b 97 None
Albumin 66 None
Ovalbumin 45 2
β-casein 30 5
Lysozyme 15 None
*Potential phosphorylation sites. Not all sites may be phosphorylated.
The PhosDecor™ Control contains six proteins two of which are phosphorylated, providing a set of positive and negative controls for use with the PhosDecor Stain (Catalog Number P7374) or with the PhosDecor Fluorescent Phosphoprotein In-Gel Detection Kit (Catalog Number PDECOR). When reconstituted the six proteins are of approximately 250 μg/ml producing SDS-PAGE bands with approximately equal total protein staining intensities. This control is not intended for use in sample quantitation.
Legal Information
PhosDecor is a trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co.