Components
Standards include:
• Myosin (205,000 Da)
• β-Galactosidase (116,000 Da)
• Phosphorylase b (97,000 Da)
• Transferrin (80,000 Da)
• Albumin, bovine (66,000 Da)
• Glutamate dehydrogenase (55,000 Da)
• Ovalbumin (45,000 Da)
• Carbonic anhydrase (30,000 Da)
• Trypsin inhibitor (21,000 Da)
• Lysozyme (14,000 Da)
• Aprotinin (6,500 Da)
Each protein is present at ~60 μg/ml
Application
Molecular Probes protein molecular weight standards include 11 proteins from 6,500 to 205,000 daltons for use as molecular weight standards in SDS-polyacrylamide gel electrophoresis.
Legal Information
SYPRO is a registered trademark of Molecular Probes, Inc.