![]() |
Synonym
2
Phenylpyruvic acid sodium salt
Molecular Formula
C6H5CH2COCOONa
Molecular Weight
186.14
CAS Number
Beilstein Registry Number
4770978
MDL number
PubChem Substance ID
EG/EC Number
204-053-2
| ||||||||