Application
Designed for chemiluminescent detection of FLAG epitope-tagged fusion protein on Western blots. The CPS peroxidase substrate (CPS-1-60) included in the kit provides high sensitivity chemiluminescent detection of less than 1 ng of FLAG-tagged protein. Immunostaining with the provided ANTI-FLAG M2 monoclonal antibody-peroxidase conjugate eliminates non-specific background and simplifies the procedure compared to the use of unconjugated anti-FLAG antibodies with anti-mouse IgG secondary antibody peroxidase conjugates.
General description
All of the components of this ProteoQwest kit have been extensively tested and optimized.
Other Notes
Suitability
This kit is designed for 12 mini-gel sized (10 × 10 cm) blots.
Legal Information
ProteoQwest is a trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co.
FLAG is a registered trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co.