Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Product No. Only)
Supplier X Reference
Substructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
R0277
Sigma
Tissue slide
from rat skeletal muscle, single organ
Expand/Collapse All
Price and Availability
Descriptions
Application
Suitable for immunohistochemistry and in situ hybridization. May be used for cellular localization of tissue specific mRNA and protein expression.
Other Notes
Each slide contains a single tissue.
Preparation Note
Paraffin embeded tissue sections, approximately 5 μm thick.
Properties
biological source
from rat skeletal muscle
Safety
WGK Germany
3
Related Categories
...
Signal Transduction Research Tools
>
Immunohistology and Blotting Controls
...
Supplementary Products
>
Tissue and Protein Controls
Scroll Up
Scroll Down
Related Information
R0277 - Datasheet (26kb)
MSDS
Certificate of Analysis
More Information
Links
Similar Products
Related Categories
Page Options
Printer Friendly View
Bulk Quote
Ask A Scientist
Last 5 Products Viewed
R0277 (Sigma)
en-us-footer US
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.