Other Notes
Supplied with 10x Restriction Endonuclease Buffer SH (B3657).
Linkage
Isoschizomers : BstN I, Apy I, BsiL I, BstO I
Other Notes
Comment: Ad-2 DNA is the substrate in the assays. EcoR II will only partially cleave DNA isolated from E. coli strains that have the dcm methylase (dcm + strains).
Specificity
Recognition sequence:5′-/CC(AT)GG-3′
Ligation and recutting results: After 2-10-fold EcoR II overdigestion of 1 μg Ad-2 DNA substrate, results in 100% cutting, >90% of fragments can be ligated, and >90% recut.
Heat inactivation: 65 °C for 10 minutes.
Physical form
Solution in 20 mM Tris-HCl, pH 7.0, 1 mM EDTA, 300 mM KCl, 1 mM dithioerythritol, 0.01% polydocanol, 50% glycerol (v/v) at 4°C