antibody form
affinity isolated antibody
form
buffered aqueous solution
mol wt
antigen mol wt ~55 kDa
species reactivity
mouse
application(s)
immunoblotting: 2-4 μg/mL using extracts of NIH3T3 cells induced with 10% fetal calf serum
immunoprecipitation: 2.5-5.0 μg using extracts of NIH3T3 cells induced with 10% fetal calf serum
indirect immunofluorescence: 4-6 μg/mL using methanol-acetone fixed sections of NIH3T3 cells induced with 10% fetal calf serum
Gene Information
shipped in
dry ice
storage temp.
−20°C