Other Notes
Supplied with 10x Restriction Endonuclease Buffer SB (B8781).
Linkage
Isoschizomer: BsoB I.
Specificity
Recognition sequence: 5′-C/PyCGPuG-3′
Ligation and recutting results: After 2-10-fold Ava I overdigestion of 1 μg λ DNA substrate, results in 100% cutting, >95% of fragments can be ligated, and >95% recut.
Heat inactivation: 65 °C for 15 minutes.
Physical form
Solution in 20 mM Tris-HCl, pH 8.0, 0.1 mM EDTA, 100 mM NaCl, 10 mM 2-Mercaptoethanol, 0.01% Polydocanol (v/v), 50% glycerol (v/v) at 4°C