Other Notes
Supplied with 10x Restriction Endonuclease Buffer SA (B7531).
Comment: Activity of Ava II blocked by overlapping dcm methylation
Specificity
Recognition sequence: 5′-G/G(A,T)CC-3′
Ligation and recutting results: After 2-10-fold Ava II overdigestion of 1 μg λ DNA substrate, results in 100%cutting, >90% of fragments can be ligated, and >95% recut.
Heat inactivation: 65 °C for 15 minutes.
Physical form
Solution in 10 mM Tris-HCl, pH 7.4, 50 mM KCl, 0.1 mM EDTA, 1 mM DTT, 0.2 mg/ml BSA, 50% glycerol (v/v) at 4°C