General description
Handheld, single-use Osborn Scientific® Biowarfare Agent Detection Devices for qualitative environmental testing for Ricin Toxin. Accurately detects minute amounts of Ricin Toxin on surfaces or in solutions in as little as 3 minutes. Tests are individually packaged in a foil pouch along with the simulant, pipette and swab. No additional accessories needed to perform an immediate evaluation, anywhere in the world.
• Detection Level - As little as 50 nanograms of Ricin Toxin on surfaces, 400 nanograms in solutions
• Sensitivity - 100% accurate as independently tested by the Department of Defense
• Specificity - Testing of substances evaluated produced no false positives or negatives
• Easy to use - Completely self contained