MISSION shRNA clones are sequence-verified shRNA lentiviral plasmids (pLKO.1-puro) provided as frozen bacterial glycerol stocks (Luria Broth, carbenicillin at 100 µg/ml and 10% glycerol) in Escherichia coli for propagation and downstream purification of the shRNA clones. pLKO.1 contains the puromycin selection marker for transient or stable transfection. In addition, the plasmids may be used to generate lentiviral particles in packaging cell lines. Orders of 5 or less target sets (<25 clones or constructs) are provided in individual 1.4 ml tubes. Orders of more than 5 target sets (>25 clones or constructs) are provided in barcoded 96-well plates with a CD containing plate map positions.