General description
MISSION siRNA Library Format:
• 21mer siRNA duplexes with dTdT overhangs
• 3 individual siRNA duplexes per target gene designed using Rosetta Inpharmatics' design algorithm
• All siRNA duplexes spotted in 96 well microplates with 80 duplexes per plate; delivered dry with a final quantity of 1 nmol per duplex
• Control siRNA sequences included on all plates
• Supplied with a CD with plate maps, target gene list and full sequence information for the siRNAs
Legal Information
MISSION is a registered trademark of Sigma-Aldrich Biotechnology LP and Sigma-Aldrich Co.