| T4685 Sigma |
Trisacryl GF 05M Grade, aqueous ethanol suspension, 40-80 μm (wet), exclusion limit 3,000 Da (calculated by experimental standard curve), fractionation range 200-2,500 Da |
|
|||||||||
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. |