|
| U8628 Sigma |
Ultrogel AcA 54aqueous ethanol suspension, 60-140 μm (wet), exclusion limit 90,000 Da, fractionation range 5,000-70,000 Da (globular proteins) |
|
|||||||
| Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. |