General description
Dispensette HF (green color-code): for dispensing hydrofluoric acid (HF). Maximum permitted concentration 52%.
• Easy to dismantle for cleaning, with replaceable filling valves
• Autoclavable at 121 °C
• Fast volume adjustment with analog slide
• Simple calibration adjustment with supplied tool
• SafetyPrime™ recirculation valve reduces reagent waste and the safety discharge system reduces the risk of inadvertent dispensing and splashes.
• To allow the bottle label to face the user, the valve block can be rotated 360°.
• A telescopic filling tube adjusts easily to different size bottles.
The parts of the Dispensette HF which are in contact with medium are made of:
ceramic, platium-iridium, ETFE, FEP & PFA. The discharge tube safety screw cap is PP. Operating limits: vapor pressure max. 500 mbar, viscosity max. 500 mm2/s, temperature max. 40 °C, density max. 3.8 g/cm3. Accuracy: ±0.5%, precision ±0.1%.
Conformity certified.
Legal Information
BRAND is a trademark of Brand GmbH & Co. KG.
Dispensette is a registered trademark of Brand GmbH & Co. KG.
SafetyPrime is a trademark of Brand GmbH & Co. KG.