Order Center
Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Prod. No. Only)
Supplier X Reference
SubStructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
Z337587
Sigma
Consort power supply accessories
Pair of 2 mm sockets
Expand/Collapse All
Price and Availability
Properties
availability
available only in EU
Related Categories
...
Manufacturer Browser
>
Consort
...
Electrophoresis Equipment
>
Power Supplies and Accessories
Scroll Up
Scroll Down
Related Information
MSDS
Certificate of Analysis
More Information
Links
Similar Products
Related Categories
Page Options
Printer Friendly View
Ask A Scientist
Email Page
Last 5 Products Viewed
Z337587 (Sigma)
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.