General description
• Quick and easy-to-use. Supplied ready assembled, saving time in the laboratory
• Prefilter versions available for difficult-to-filter samples
• Frosted area on tube for easy sample identification
• 400μL insert capacity (receiving tube capacity with insert: 1.25mL; without insert: 2.0mL)
Applications:-
• Removal of cells from culture media
• Particle removal from solvents
• Liquid chromatography sample preparation
• Removal of bacteria from sample material
• Fractionation/purification of proteins