General description
All Acculab balances listed below
• tare over full capacity
• have keypad auto-calibration function
• have auto-off with memory to prolong battery life
• have four weighing modes (g, avoirdupois oz, troy oz, dwt), plus parts counting on V-400, V-600, V-1200, and V-2400 models
• LCD has overload/underload indicators and low battery indicator
• operate with fully compensated strain-gauge transducer technology, optimal between 18-25 °C
• AC adapter included
General-purpose laboratory balances with a wide range of capacities. Excellent value in precision weighing.
Legal Information
Acculab is a registered trademark of Globaltec Corp.