|
| Z360473 Sigma |
Utility boxes, plasticL × W × H 129 mm × 75 mm × 105 mm |
|
|||||||
| Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. |