General description
SigmaScan Pro's comprehensive list of image types ensures image compatibility- including up to 16-bit grayscale images. You can convert images to different types, paste or splice images from one type to another, or modify pixel resolution while maintaining the aspect ration of the image. The powerful Histogram Stretch option lets you sharpen the contrast of color or grayscale images. Other enhancement techniques allow you to correct background lighting effects, apply color templates to grayscale images for highlighting hidden features or employ mathematical filters to add, remove, erode or dilate objects on an image. Plus, SigmaScan Pro's Image Math functions let you remove noise and highlight features by adding, subtracting or averaging the pixel intensities of different images.
System Requirements
• Pentium processor running Windows 95, 98, 2000 or NT 4
• 16MB RAM (64MB recommended)
• SVGA monitor, 800x600 resolution 256 colors (True Color recommended)
• 15MB hard disk space
Legal Information
SigmaScan is a registered trademark of Jandel Scientific.