General description
Designs buffers using the Henderson-Hasselbalch equation. Has a library of over 500 buffer recipes! Also will calculate % constituent, ionic strength, dilution values, standardization values, pH, pK, and pKa.
System Requirements
400 MHz or faster with 128 MB RAM running Windows 9x, Me, NT, 2000, or XP
50MB minimum available HDD space for most application directories and 1 GB if a database program allowing more for growth
File Server running Windows NT or Windows 2000, using Windows or Novell (for network installations).
Legal Information
Windows is a registered trademark of Microsoft Corp.