General description
Includes new features like exploratory graphs, a graph editor and an enhanced report editor.
Powerful statistical analysis made easy. Analyze your data by almost any standard statistical method you choose. Print detailed reports of results or enhance your reports with graphs produced by SigmaPlot.
SigmaStat ensures that you:
• Use the proper statistical method to analyze your data
• Avoid the risk of statistical error
• Interpret the results correctly
• Generate an appropriate display and a professional report
·
Be sure you've analyzed your data correctly. And save time, too!
SigmaStat is used by thousands of research scientists worldwide to:
• Compare effects among groups, including before-and-after or repeated measures studies
• Conduct survival and reliability analyses
• Analyze rates and proportions
• Perform regression and correlation analyses
• Calculate power and sample size
System Requirements
Windows 98, 2000, NT or XP; Pentium 200 or higher
Legal Information
SigmaStat is a registered trademark of Jandel Scientific.