General description
When pressed against a microscope slide, forms a sealed chamber for thick and free-floating specimens. No compression artifacts like those caused by ordinary cover slips. Ideal for confocal and image analysis by light or fluorescence microscopy. Chambers adhere to wet or dry surfaces and can be removed and re-applied without damage to specimens. Peel-and-stick adhesive versions form leak-proof enclosures for permanent mounting. Designed to accommodate 25 mm filters for particle analysis and microbiological examination. Apply to 50 × 75 mm glass slide.
Legal Information
CoverWell is a trademark of Grace Bio-Labs, Inc.