Order Center
Product Name or No.
Keyword (Full Text)
MISSION shRNA-RNAi
MISSION siRNA
Molecular Formula
CAS No.
MDL No.
MSDS (Prod. No. Only)
Supplier X Reference
SubStructure
YFG Search
Product Name or No.
Keyword (fulltext)
Molecular Formula
CAS No.
MDL No.
Enzyme Commission (EC) Number
EG/EC No.
FEMA No.
Beilstein Registry No.
Color Index
Product Name
Product No.
Product Category
Chromatography Application
Supplier Part Number
MISSION shRNA-RNAi
MISSION siRNA
Z365963
Sigma
LIFEGUARD culture boxes
Elastic sealing bands
Expand/Collapse All
Price and Availability
Identifiers
Synonym
Elastic Sealing Bands for LIFEGUARD Culture boxes
Properties
material
rubber
description
Autoclavable
Related Categories
...
Culture Boxes and Accessories
>
Membrane Rafts, Culture Boxes , and Accessories
...
Cell Culture Supplies
>
Plant Cell culture supplies
Scroll Up
Scroll Down
Related Information
Plant Biotech Guide (254kb)
MSDS
Certificate of Analysis
More Information
Links
Similar Products
Related Categories
Page Options
Printer Friendly View
Bulk Quote
Ask A Scientist
Email Page
Last 5 Products Viewed
Z365963 (Sigma)
Site Use Terms
|
Terms and Conditions of Sale
|
Privacy
|
Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc.