General description
Counting colonies and plaques on culture plates is simple, accurate, and easy on the eyes. The pen-style counter marks, counts, and beeps to confirm each touch. Select count-up, count-down, or error-correction mode. Felt tip markers (one red and one black provided) make water-resistant marks on glass and plastic; alcohol removes marks. The mini light box has a removable grid to assist counting. The free-standing 1.75× mini magnifier enhances the view while it leaves both hands free. All components can be ordered separately and have their own independent laboratory applications.
Legal Information
Scienceware is a registered trademark of Bel-Art Products.