|
| Z368407 Sigma |
Disposable dialyzersMWCO 50,000 Da, capacity 2 mL, cellulose acetate/cellulose nitrate mixed esters membrane |
|
|||||||
| Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. |