|
| Z368482 Sigma |
Disposable dialyzersMWCO 8,000 Da, capacity 2 mL, regenerated cellulose membrane |
|
|||||||
| Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. |