General description
A reversible motor rotates rack of disposable dialyzers at 10 rpm, agitating sample and buffer for rapid dialysis with minimum buffer. Top plate has holes to accommodate twenty 0.5, 1 or 2 mL and twenty-four 5 mL dialyzers; positions are numbered for sample identification. Unit comes with one of each dialyzer listed above.
A reversible motor rotates rack of disposable dialyzers at 10rpm, agitating sample and buffer for rapid dialysis with minimum buffer. Top plate has holes to accommodate twenty 0.5, 1, or 2-ml and twenty-four 5-ml dialyzers; positions are numbered for sample identification.