|
| Z373230 Sigma |
Ohaus® Explorer® balance62 g weighing capacity, resolution: 0.1 mg, AC input 220 V, internal calibration |
|
|||||||
| Copyrights © 2008 Sigma-Aldrich Co. All Rights Reserved. Reproduction of any materials from the site is strictly forbidden without permission. Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. |