General description
This fully updated edition pulls together diverse areas and applications of CE; the result is a volume that serves scientists in biotechnology, clinical, and pharmaceutical chemistry. The clear organization distinguishes general information − theory, modes of CE, detection, and data analysis − from specifics − with chapters on small molecules, peptides, proteins, carbohydrates, and nucleic acids as analytes. Some of the most useful chapters explore highly specialized applications of CE to large-scale DNA sequencing, drug analysis in biological fluids, binding studies, and enzyme assays.