General description
These clip-on units will convert an ordinary water bath into a precision temperature-controlled bath. Each incorporates a thermostatted heater unit, circulating pump, connectors for optional external circulation, and an adjustable over-temperature cutoff. Refrigeration required for operation below ambient (cooling coil available as an option if not supplied standard).
All submerged parts are made of stainless steel or high-temperature, self-extinguishing plastic.
Supplied as circulator only (order bath separately).
Thermoregulator bridge mounting plate recommended for use above 150 °C.
Legal Information
Techne is a registered trademark of Techne, Inc.