General description
Five different models offer a selection of temperature ranges and features, and accept a wide variety of insert blocks for your specific needs. All are sturdily constructed of aluminum and engineering-grade plastic with all solid-state controls, and are provided with a block removal tool. Efficient insulation keeps case cool even at maximum operating temperature. Pilot lights indicate power on/off, heater action, and over-temperature warning.
Accepts up to two insert blocks; Temperature range: 25-105 °C; Setting is by three adjustable presets.
Legal Information
Techne is a registered trademark of Techne, Inc.
Dri-Block is a registered trademark of Techne, Inc.