Application
The patented UNIFILTER products are convenient, ready to use, disposable filter bottom microplates. Available in 96 well formats, the UNIFILTER microplates offer a choice of filter media that work well with the most stringent applications. UNIFILTERS can be used to process 96 samples at a time. The unique drip director design allows for analysis of the filtrate or the retentate. UNIFILTERS are available in many different variations of well formats and range from 100 μL to 2mL, using a wide variety of filtration media.
General description
• Patented design prevents well to well crosstalk.
• Large variety of well volumes economizes on expensive consumables.
• Choice of filter media allows control of flow rates and retention characteristics.
• Standard overall dimensions fit most plate readers, auto samplers, robots.
Legal Information
Whatman is a registered trademark of Whatman International Ltd.
UNIFILTER is a registered trademark of Whatman International Ltd.